2-Methyl-2-(4-methylphenyl)morpholine 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Methyl-2-(p-tolyl)morpholine
H50732.03
H50732.MD
H50732.03EA
9720
CZK
H50732.03
H50732.MD
2-Methyl-2-(4-methylphenyl)morpholine 97%
2-Methyl-2-(4-methylphenyl)morpholine
Purity: 97%
CAS No.: 902836-81-1
Molecular Formula: C12H17NO
Formula Weight: 191.28
Melting Point: 49-51°
MDL No.: MFCD08060961
Vzorec:
C₁₂H₁₇NO MW: 191,28 g/mol Bod tání: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD08060961 CAS číslo: 902836-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...