3-(2-Oxocyclohexyl)propanenitrile 99%
Dodavatel: Thermo Scientific
|
Varování
|
H50722.03
H50722.06
H50722.03EA
1580
CZK
H50722.03
H50722.06
3-(2-Oxocyclohexyl)propanenitrile 99%
3-(2-Oxocyclohexyl)propanenitrile
Purity: 99%
CAS No.: 4594-78-9
Molecular Formula: C9H13NO
Formula Weight: 151.21
Boiling Point: 138-142°/10mm
UN No.: UN3276
MDL No.: MFCD00013797
Vzorec:
C₉H₁₃NO MW: 151,21 g/mol Bod varu: 138…142 °C (10 mmHg) Hustota: 1,021 Storage Temperature: Ambient |
MDL Number:
MFCD00013797 CAS číslo: 4594-78-9 EINECS: 224-991-6 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...