Tisk…

8-hydroxyquinoline-2-carbonitrile 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H50433.03 H50433.06
H50433.03EA 2400 CZK
H50433.03 H50433.06
8-hydroxyquinoline-2-carbonitrile 98%
8-hydroxyquinoline-2-carbonitrile

Purity: 98% CAS No.: 6759-78-0 Molecular Formula: C10H6N2O Formula Weight: 170.17 Melting Point: 133-136° UN No.: UN3439 MDL No.: MFCD00216717

Vzorec: C₁₀H₆N₂O
Bod tání: 133…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00216717
CAS číslo: 6759-78-0
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR