4-(Diethylamino)butylamine 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
N1,N1-Diethylbutane-1,4-diamine
H50278.03
H50278.06
H50278.03EA
3130
CZK
H50278.03
H50278.06
4-(Diethylamino)butylamine 96%
4-(Diethylamino)butylamine
Purity: 96%
CAS No.: 27431-62-5
Molecular Formula: C8H20N2
Formula Weight: 144.26
Boiling Point: 72-24°/10mm
UN No.: UN2735
MDL No.: MFCD00014836
Vzorec:
C₈H₂₀N₂ MW: 144,26 g/mol Bod varu: 72…24 °C (10 mmHg) Hustota: 0,83 Storage Temperature: Ambient |
MDL Number:
MFCD00014836 CAS číslo: 27431-62-5 EINECS: 248-456-1 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...