Tisk…

N-Methyl-2-phenoxyethanamine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H50258.03 H50258.MD
H50258.03EA 8480 CZK
H50258.03 H50258.MD
N-Methyl-2-phenoxyethanamine 98%
N-Methyl-2-phenoxyethanamine

Purity: 98% CAS No.: 37421-04-8 Molecular Formula: C9H13NO Formula Weight: 151.21 UN No.: UN2735 MDL No.: MFCD00085690

Vzorec: C₉H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00085690
CAS číslo: 37421-04-8
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR