Tisk…

3-(2-Methoxy-3-pyridyl)acrylic acid 95%

Dodavatel: Thermo Fisher Scientific
Varování

H50063.03 H50063.06
H50063.03EA 4330 CZK
H50063.03 H50063.06
3-(2-Methoxy-3-pyridyl)acrylic acid 95%
3-(2-Methoxy-3-pyridyl)acrylic acid

Purity: 95% CAS No.: 131674-41-4 Molecular Formula: C9H9NO3 Formula Weight: 179.18 MDL No.: MFCD07781156

Vzorec: C₉H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD07781156
CAS číslo: 131674-41-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR