Tisk…
Platina 5% on activated carbon paste Type 128M
Dodavatel: Thermo Fisher Scientific
H37641.06 H37641.14 H37641.22
H37641.06EA 3470 CZK
H37641.06 H37641.14 H37641.22
Platina 5% on activated carbon paste Type 128M
Platina
CAS No.: 7440-06-4 Physical Form: Solid Appearance: Black Odour: Odourless MDL No.: MFCD00011179
Vzorec: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1