Tisk…
Paladium 5% on activated carbon Type 487
Dodavatel: Thermo Fisher Scientific
Nebezpečí
H37587.06 H37587.14
H37587.06EA 2840 CZK
H37587.06 H37587.14
Paladium 5% on activated carbon Type 487
Palladium
CAS No.: 7440-05-3 Physical Form: Powder Appearance: Dark grey Odour: Odourless UN No.: UN1362 MDL No.: MFCD03457879
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
EINECS: 231-115-6
UN: 1362
ADR: 4.2 III