1,3-Dibromoacetone 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H37325.14
H37325.06
H37325.14EA
10610
CZK
H37325.14
H37325.06
1,3-Dibromoacetone 98%
1,3-Dibromoacetone
Purity: 98%
CAS No.: 816-39-7
Molecular Formula: C3H4Br2O
Formula Weight: 215.87
Storage Temperature: 2°C to 8°C
Flash Point: 102°(216°F)
Physical Form: Crystals or fused solid
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Melting Point: 29-30°
Boiling Point: 97-98°/21mm
Beilstein Reference: 1740389
UN No.: UN1759
MDL No.: MFCD00013540
Vzorec:
BrH₂CC(O)CH₂Br Bod varu: 97…98 °C (21 mmHg) Bod tání: 29…30 °C Bod vzplanutí: 102 °C (216 °F) |
MDL Number:
MFCD00013540 CAS číslo: 816-39-7 EINECS: 212-430-8 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...