Tisk…
Platina 10% on activated carbon paste Type 128M
Dodavatel: Thermo Fisher Scientific
Synonyms: Platinium
H36044.06 H36044.14 H36044.22
H36044.06EA 7210 CZK
H36044.06 H36044.14 H36044.22
Platina 10% on activated carbon paste Type 128M
Platina
CAS No.: 7440-06-4 Physical Form: Solid Appearance: Black Odour: Odourless MDL No.: MFCD00011179
Vzorec: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
UN: 3088
ADR: 4.2,III