Tisk…

1-Methyl-3-indolemethylamine 96%

Dodavatel: Thermo Fisher Scientific
Varování

H34418.MD H34418.03 H34418.06
H34418.MDEA 1540 CZK
H34418.MD H34418.03 H34418.06
1-Methyl-3-indolemethylamine 96%
1-Methyl-3-indolemethylamine

Purity: 96% CAS No.: 19293-60-8 Molecular Formula: C10H12N2 Formula Weight: 160.22 MDL No.: MFCD06657101

Vzorec: C₁₀H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD06657101
CAS číslo: 19293-60-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR