3-(2-Bromophenyl)-1H-pyrazole 97%
Dodavatel: Thermo Fisher Scientific
H34406.03
H34406.09
H34406.03EA
1210
CZK
H34406.03
H34406.09
3-(2-Bromophenyl)-1H-pyrazole 97%
3-(2-Bromophenyl)-1H-pyrazole
Purity: 97%
CAS No.: 114382-20-6
Molecular Formula: C9H7BrN2
Formula Weight: 223.07
Melting Point: 130-132°
MDL No.: MFCD01940435
Vzorec:
C₉H₇BrN₂ Bod tání: 130…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD01940435 CAS číslo: 114382-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...