4-(2-Thienyl)benzenesulphonyl chloride 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H34399.MD
H34399.03
H34399.06
H34399.MDEA
1140
CZK
H34399.MD
H34399.03
H34399.06
4-(2-Thienyl)benzenesulphonyl chloride 96%
4-(2-Thienyl)benzenesulphonyl chloride
Purity: 96%
CAS No.: 209592-91-6
Molecular Formula: C10H7ClO2S2
Formula Weight: 258.75
Melting Point: 107-108°
UN No.: UN3261
MDL No.: MFCD11655569
Vzorec:
C₁₀H₇ClO₂S₂ MW: 258,75 g/mol Bod tání: 107…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD11655569 CAS číslo: 209592-91-6 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...