2-Amino-5-iodonicotinonitrile 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H34191.03
H34191.06
H34191.03EA
2700
CZK
H34191.03
H34191.06
2-Amino-5-iodonicotinonitrile 97%
2-Amino-5-iodonicotinonitrile
Purity: 97%
CAS No.: 1347815-41-1
Molecular Formula: C6H4IN3
Formula Weight: 245.02
Melting Point: 193-197°
UN No.: UN3439
MDL No.: MFCD20040120
Vzorec:
C₆H₄IN₃ Bod tání: 193…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD20040120 CAS číslo: 1347815-41-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...