Tisk…

[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4'-Chloro-[1,1'-biphenyl]-4-sulphonyl chloride, 4'-Chlorobiphenyl-4-sulphonyl chloride

H34182.09 H34182.03
H34182.09EA 8310 CZK
H34182.09 H34182.03
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride

Purity: 96% CAS No.: 20443-74-7 Molecular Formula: C12H8Cl2O2S Formula Weight: 287.17 Melting Point: 107-108° UN No.: UN3261 MDL No.: MFCD01631918

Vzorec: C₁₂H₈Cl₂O₂S
MW: 287,17 g/mol
Bod tání: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01631918
CAS číslo: 20443-74-7
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR