Tisk…

3-(4-Formylphenyl)propionic acid 96%

Dodavatel: Thermo Scientific
Varování

H33873.MD H33873.03 H33873.06
H33873.MDEA 1690 CZK
H33873.MD H33873.03 H33873.06
3-(4-Formylphenyl)propionic acid 96%
3-(4-Formylphenyl)propionic acid

Purity: 96% CAS No.: 34961-64-3 Molecular Formula: C10H10O3 Formula Weight: 178.19 Melting Point: ca 136° dec MDL No.: MFCD08234824

Vzorec: C₁₀H₁₀O₃
MW: 178,19 g/mol
Bod tání: ∼ 136 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD08234824
CAS číslo: 34961-64-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR