Tisk…

1-Chloromethyl-1H-1,2,4-triazole hydrochloride 96%

Dodavatel: Thermo Scientific
Varování

H33830.03 H33830.06
H33830.03EA 2920 CZK
H33830.03 H33830.06
1-Chloromethyl-1H-1,2,4-triazole hydrochloride 96%
1-Chloromethyl-1H-1,2,4-triazole hydrochloride

Purity: 96% CAS No.: 84387-62-2 Molecular Formula: C3H4ClN3.HCl Formula Weight: 154.00 Melting Point: 138-145° MDL No.: MFCD00964947

Vzorec: C₃H₄ClN₃·HCl
MW: 154,00 g/mol
Bod tání: 138…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00964947
CAS číslo: 84387-61-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR