2-Methylbenzothiazol-6-ol 96%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Methylbenzo[d]thiazol-6-ol
H33812.03
H33812.06
H33812.03EA
3290
CZK
H33812.03
H33812.06
2-Methylbenzothiazol-6-ol 96%
2-Methylbenzothiazol-6-ol
Purity: 96%
CAS No.: 68867-18-5
Molecular Formula: C8H7NOS
Formula Weight: 165.21
Melting Point: 159-161°
MDL No.: MFCD00227234
Vzorec:
C₈H₇NOS Bod tání: 159…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00227234 CAS číslo: 68867-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...