Tisk…

3-(4-Nitrophenyl)propionic acid 96%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Nitrohydrocinnamic Acid, 3-(4-Nitrophenyl)propanoic acid

H33663.09 H33663.18
H33663.09EA 3540 CZK
H33663.09 H33663.18
3-(4-Nitrophenyl)propionic acid 96%
3-(4-Nitrophenyl)propionic acid

Purity: 96% CAS No.: 16642-79-8 Molecular Formula: C9H9NO4 Formula Weight: 195.17 Melting Point: 164-168° MDL No.: MFCD00126834

Vzorec: C₉H₉NO₄
MW: 195,17 g/mol
Bod tání: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00126834
CAS číslo: 16642-79-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR