Tisk…

3-(2-Chloro-6-fluorophenyl)propionic acid 96%

Dodavatel: Thermo Fisher Scientific
Varování

H33586.03 H33586.06
H33586.03EA 2480 CZK
H33586.03 H33586.06
3-(2-Chloro-6-fluorophenyl)propionic acid 96%
3-(2-Chloro-6-fluorophenyl)propionic acid

Purity: 96% CAS No.: 88740-77-6 Molecular Formula: C9H8ClFO2 Formula Weight: 202.61 Melting Point: 59-63° MDL No.: MFCD06660331

Vzorec: C₉H₈ClFO₂
Bod tání: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD06660331
CAS číslo: 88740-77-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR