5-Bromo-3-iodo-2-hydroxypyridine 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Bromo-3-iodopyridin-2(1H)-one, 5-Bromo-2-hydroxy-3-iodopyridine
H33495.03
H33495.06
H33495.14
H33495.03EA
683
CZK
H33495.03
H33495.06
H33495.14
5-Bromo-3-iodo-2-hydroxypyridine 97%
5-Bromo-3-iodo-2-hydroxypyridine
Purity: 97%
CAS No.: 381233-75-6
Molecular Formula: C5H3BrINO
Formula Weight: 299.89
Melting Point: 226-227°
MDL No.: MFCD08688586
Vzorec:
C₅H₃BrINO MW: 299,89 g/mol Bod tání: 226…227 °C Storage Temperature: Ambient |
MDL Number:
MFCD08688586 CAS číslo: 381233-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...