3,3,3-Trifluoro-DL-alanine 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H33461.MD
H33461.03
H33461.MDEA
5560
CZK
H33461.MD
H33461.03
3,3,3-Trifluoro-DL-alanine 97%
3,3,3-Trifluoro-DL-alanine
Purity: 97%
CAS No.: 17463-43-3
Molecular Formula: C3H4F3NO2
Formula Weight: 143.06
Melting Point: 210° dec.
MDL No.: MFCD00004263
Vzorec:
C₃H₄F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00004263 CAS číslo: 17463-43-3 EINECS: 241-478-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...