Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Dodavatel: Thermo Scientific
Synonyms:
Pyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Methyl ester, 1-Boc-3-pyrrolidinecarboxylic acid methyll ester, 1-tert-Butyl 3-Methyl Pyrrolidine-1,3-dicarboxylate, 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid methyll ester, Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate, Methyl 1-Boc-3-pyrrolidinecarboxylate
H33407.03
H33407.06
H33407.03EA
3000
CZK
H33407.03
H33407.06
Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Methyl-1-boc-3-pyrrolidinecarboxylate
Purity: 97%
CAS No.: 122684-33-7
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Boiling Point: 105°/0.2mm
MDL No.: MFCD04038683
Vzorec:
C₁₁H₁₉NO₄ MW: 229,28 g/mol Bod varu: 105 °C (0,2 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD04038683 CAS číslo: 122684-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...