1,3-benzothiazol-5-amine 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole
H33309.03
H33309.06
H33309.03EA
2640
CZK
H33309.03
H33309.06
1,3-benzothiazol-5-amine 96%
1,3-benzothiazol-5-amine
Purity: 96%
CAS No.: 1123-93-9
Molecular Formula: C7H6N2S
Formula Weight: 150.20
Melting Point: 55-59°
MDL No.: MFCD04115282
Vzorec:
C₇H₆N₂S MW: 150,199 g/mol Bod tání: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115282 CAS číslo: 1123-93-9 EINECS: 214-381-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...