Tisk…

6-Chloro-2-iodo-3-hydroxypyridine 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 6-Chloro-2-iodopyridin-3-ol

H33127.03 H33127.09
H33127.03EA 2210 CZK
H33127.03 H33127.09
6-Chloro-2-iodo-3-hydroxypyridine 98%
6-Chloro-2-iodo-3-hydroxypyridine

Purity: 98% CAS No.: 188057-26-3 Molecular Formula: C5H3ClIO Formula Weight: 255.44 Melting Point: 191-193° MDL No.: MFCD07636749

Vzorec: C₅H₃ClIO
Bod tání: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD07636749
CAS číslo: 188057-26-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR