Tisk…

3-Pyrrol-1-ylbenzoic acid 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-(1H-Pyrrol-1-yl)benzoic acid

H33124.03 H33124.09
H33124.03EA 2940 CZK
H33124.03 H33124.09
3-Pyrrol-1-ylbenzoic acid 97%
3-Pyrrol-1-ylbenzoic acid

Purity: 97% CAS No.: 61471-45-2 Molecular Formula: C11H9NO2 Formula Weight: 187.20 Melting Point: 175-178° MDL No.: MFCD02656610

Vzorec: C₁₁H₉NO₂
MW: 187,2 g/mol
Bod tání: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD02656610
CAS číslo: 61471-45-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR