1-Methylpiperidine-4-carboxylic acid hydrochloride 96%
Dodavatel: Thermo Scientific
|
Varování
|
H33083.03
H33083.06
H33083.03EA
1740
CZK
H33083.03
H33083.06
1-Methylpiperidine-4-carboxylic acid hydrochloride 96%
1-Methylpiperidine-4-carboxylic acid hydrochloride
Purity: 96%
Molecular Formula: C7H13NO2.HCl
Formula Weight: 179.65
Melting Point: 225°
MDL No.: MFCD00190235
Vzorec:
C₇H₁₄ClNO₂ Bod tání: 225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190235 CAS číslo: 71985-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...