4-(1,3-Benzothiazol-2-yl)phenylamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-(2-Benzothiazolyl)aniline, 4-(Benzo[d]thiazol-2-yl)aniline, 2-(4-Aminophenyl)benzothiazole
H33052.03
H33052.06
H33052.03EA
3680
CZK
H33052.03
H33052.06
4-(1,3-Benzothiazol-2-yl)phenylamine 98%
4-(1,3-Benzothiazol-2-yl)phenylamine
Purity: 98%
CAS No.: 6278-73-5
Molecular Formula: C13H10N2S
Formula Weight: 226.30
Melting Point: 152-154°
MDL No.: MFCD00182896
Vzorec:
C₁₃H₁₀N₂S MW: 226,3 g/mol Bod tání: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00182896 CAS číslo: 6278-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...