2,2'-O-Cyclouridine 98%
Dodavatel: Thermo Scientific
Synonyms:
2,2'-O-Anhydro-(1-beta-D-arabinofuranosyl)uracil, (2R,3R,3aS,9aR)-3-Hydroxy-2-(hydroxymethyl)-2,3,3a,9a-tetrahydro-6H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-6-one, 2,2'-Anhydrouridine
H33038.03
H33038.06
H33038.03EA
968
CZK
H33038.03
H33038.06
2,2'-O-Cyclouridine 98%
2,2'-O-Cyclouridine
Purity: 98%
CAS No.: 3736-77-4
Molecular Formula: C9H10N2O5
Formula Weight: 226.19
Storage Temperature: -30°C to -10°C
Melting Point: 248-251°
Beilstein Reference: 27348885
Optical Rotation: -20° (c-4 in water)
MDL No.: MFCD00004945
Vzorec:
C₉H₁₀N₂O₅ MW: 226,19 g/mol Bod tání: 248…251 °C |
MDL Number:
MFCD00004945 CAS číslo: 3736-77-4 EINECS: 223-107-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...