Ethyl-1,3-benzothiazole-2-carboxylate 98%
Dodavatel: Thermo Scientific
Synonyms:
Benzothiazole-2-carboxylic acid ethyl ester, Ethyl 1,3-benzothiazole-2-carboxylate, Ethyl Benzothiazole-2-carboxylate
H32904.06
H32904.14
H32904.06EA
2050
CZK
H32904.06
H32904.14
Ethyl-1,3-benzothiazole-2-carboxylate 98%
Ethyl-1,3-benzothiazole-2-carboxylate
Purity: 98%
CAS No.: 32137-76-1
Molecular Formula: C10H9NO2S
Formula Weight: 207.25
Melting Point: 68-72°
MDL No.: MFCD00848360
Vzorec:
C₁₀H₉NO₂S MW: 207,25 g/mol Bod tání: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00848360 CAS číslo: 32137-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...