3-Chloro-4-nitroaniline 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Amino-2-chloronitrobenzene
H32581.03
H32581.09
H32581.03EA
755
CZK
H32581.03
H32581.09
3-Chloro-4-nitroaniline 95%
3-Chloro-4-nitroaniline
Purity: 95%
CAS No.: 825-41-2
Molecular Formula: C6H5ClN2O2
Formula Weight: 172.57
Melting Point: 157-161°
UN No.: UN2811
MDL No.: MFCD00085922
Vzorec:
C₆H₅ClN₂O₂ MW: 172.57 g/mol Bod tání: 157…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085922 CAS číslo: 825-41-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...