Tisk…

N-Boc-ethylamine 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: tert-Butyl N-ethylcarbamate, tert-Butyl ethylcarbamate

H32575.03 H32575.06
H32575.03EA 2050 CZK
H32575.03 H32575.06
N-Boc-ethylamine 97%
N-Boc-ethylamine

Purity: 97% CAS No.: 38267-76-4 Molecular Formula: C7H15NO2 Formula Weight: 145.20 Melting Point: 38-41° MDL No.: MFCD12923020

Vzorec: C₇H₁₅NO₂
Bod tání: 38…41 °C
Storage Temperature: Ambient
MDL Number: MFCD12923020
CAS číslo: 38267-76-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR