Tisk…

3-Phenyl-1H-pyrazole-4-carboxylic acid 97%

Dodavatel: Thermo Scientific
Varování

H32560.MD H32560.03 H32560.06
H32560.MDEA 1610 CZK
H32560.MD H32560.03 H32560.06
3-Phenyl-1H-pyrazole-4-carboxylic acid 97%
3-Phenyl-1H-pyrazole-4-carboxylic acid

Purity: 97% CAS No.: 5504-65-4 Molecular Formula: C10H8N2O2 Formula Weight: 188.19 Melting Point: 280-285° MDL No.: MFCD06798067

Vzorec: C₁₀H₈N₂O₂
MW: 188,19 g/mol
Bod tání: 280…285 °C
Storage Temperature: Ambient
MDL Number: MFCD03834501
CAS číslo: 5504-65-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR