2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H32544.03
H32544.09
H32544.03EA
1220
CZK
H32544.03
H32544.09
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 97%
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Purity: 97%
CAS No.: 4651-91-6
Molecular Formula: C9H10N2S
Formula Weight: 178.26
Melting Point: 141-144°
MDL No.: MFCD00128278
Vzorec:
C₉H₁₀N₂S Bod tání: 141…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128278 CAS číslo: 4651-91-6 EINECS: 225-085-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...