2-Chloro-N-cyclopropylacetamide 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H32456.09
H32456.18
H32456.09EA
1070
CZK
H32456.09
H32456.18
2-Chloro-N-cyclopropylacetamide 97%
2-Chloro-N-cyclopropylacetamide
Purity: 97%
CAS No.: 19047-31-5
Molecular Formula: C5H8ClNO
Formula Weight: 133.58
Melting Point: 80-84°
MDL No.: MFCD00276391
Vzorec:
C₅H₈ClNO Bod tání: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00276391 CAS číslo: 19047-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...