4-(Trifluoromethylsulphony)phenylacetic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H32295.03
H32295.06
H32295.03EA
3610
CZK
H32295.03
H32295.06
4-(Trifluoromethylsulphony)phenylacetic acid 98%
4-(Trifluoromethylsulphony)phenylacetic acid
Purity: 98%
Molecular Formula: C9H7F3O4S
Formula Weight: 268.21
Melting Point: 125-128°
MDL No.: MFCD04973012
Vzorec:
C₉H₇F₃O₄S MW: 268.21 g/mol Bod tání: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973012 CAS číslo: 1099597-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...