Stains-all 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H32127.03
H32127.06
H32127.03EA
2910
CZK
H32127.03
H32127.06
Stains-all 96%
Stains-all
Purity: 96%
CAS No.: 7423-31-6
Molecular Formula: C30H27BrN2S2
Formula Weight: 559.58
Beilstein Reference: 3865213
MDL No.: MFCD00078788
Vzorec:
C₃₀H₂₇BrN₂S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00078788 CAS číslo: 7423-31-6 EINECS: 231-047-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...