Tisk…

2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

Dodavatel: Thermo Scientific
Nebezpečí

H32059.03 H32059.06
H32059.03EA 2250 CZK
H32059.03 H32059.06
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile

CAS No.: 70291-62-2 Molecular Formula: C8H8N2S Formula Weight: 164.23 Physical Form: Crystalline solid Appearance: Pale brown Melting Point: 153-154° UN No.: UN3439 MDL No.: MFCD00121495

Vzorec: C₈H₈N₂S
MW: 164,23 g/mol
Bod tání: 153…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00121495
CAS číslo: 70291-62-2
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR