Tisk…

6-Bromo-2-chlorobenzothiazole 97%

Dodavatel: Thermo Scientific
Nebezpečí

H31962.03 H31962.06
H31962.03EA 1220 CZK
H31962.03 H31962.06
6-Bromo-2-chlorobenzothiazole 97%
6-Bromo-2-chlorobenzothiazole

Purity: 97% CAS No.: 80945-86-4 Molecular Formula: C7H3BrClNS Formula Weight: 248.53 Melting Point: 98-100° MDL No.: MFCD04971822

Vzorec: C₇H₃BrClNS
MW: 248,53 g/mol
Bod tání: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD04971822
CAS číslo: 80945-86-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR