Tisk…

1,3-Benzothiazole-6-carboxylic acid 96%

Dodavatel: Thermo Scientific
Varování

Synonyms: Benzo[d]thiazole-6-carboxylic acid, Benzothiazole-6-carboxylic acid

H31868.03 H31868.06 H31868.14
H31868.03EA 1540 CZK
H31868.03 H31868.06 H31868.14
1,3-Benzothiazole-6-carboxylic acid 96%
1,3-Benzothiazole-6-carboxylic acid

Purity: 96% CAS No.: 3622-35-3 Molecular Formula: C8H5NO2S Formula Weight: 179.20 Melting Point: 245-251° MDL No.: MFCD00111651

Vzorec: C₈H₅NO₂S
MW: 179,2 g/mol
Bod tání: 245…251 °C
Storage Temperature: Ambient
MDL Number: MFCD00111651
CAS číslo: 3622-35-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR