Tisk…

8-Hydroxyjulolidine-9-carboxaldehyde 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 9-Formyl-8-hydroxyjulolidine, 2,3,6,7-Tetrahydro-8-hydroxy-1H,5H-benzo[ij]quinolizine-9-carboxaldehyde, 8-Hydroxy-1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinoline-9-carbaldehyde

H31782.03 H31782.06
H31782.03EA 1970 CZK
H31782.03 H31782.06
8-Hydroxyjulolidine-9-carboxaldehyde 97%
8-Hydroxyjulolidine-9-carboxaldehyde

Purity: 97% CAS No.: 63149-33-7 Molecular Formula: C13H15NO2 Formula Weight: 217.27 Melting Point: 72-74° MDL No.: MFCD00192477

Vzorec: C₁₃H₁₅NO₂
MW: 217,27 g/mol
Bod tání: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00192477
CAS číslo: 63149-33-7
EINECS: 263-966-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR