Tisk…

2-Vinylbenzoic acid 96%

Dodavatel: Thermo Fisher Scientific
Varování

H31723.03 H31723.06
H31723.03EA 2480 CZK
H31723.03 H31723.06
2-Vinylbenzoic acid 96%
2-Vinylbenzoic acid

Purity: 96% CAS No.: 27326-43-8 Molecular Formula: C9H8O2 Formula Weight: 148.16 Storage Temperature: 2°C to 8°C Melting Point: 94° MDL No.: MFCD02066273

Vzorec: C₉H₈O₂
Bod tání: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02066273
CAS číslo: 27326-43-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR