Tisk…

2-Aminobenzo[d]thiazole-6-carboxylic acid 95%

Dodavatel: Thermo Fisher Scientific
Varování

H31512.03 H31512.06
H31512.03EA 2240 CZK
H31512.03 H31512.06
2-Aminobenzo[d]thiazole-6-carboxylic acid 95%
2-Aminobenzo[d]thiazole-6-carboxylic acid

Purity: 95% CAS No.: 93-85-6 Molecular Formula: C8H6N2O2S Formula Weight: 194.21 Melting Point: 300° MDL No.: MFCD00054180

Vzorec: C₈H₆N₂O₂S
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00054180
CAS číslo: 93-85-6
EINECS: 202-283-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR