Monomethyl suberate 98%
Dodavatel: Thermo Fisher Scientific
H30325.03
H30325.06
H30325.14
H30325.03EA
1720
CZK
H30325.03
H30325.06
H30325.14
Monomethyl suberate 98%
Monomethyl suberate
Purity: 98%
CAS No.: 3946-32-5
Molecular Formula: C9H16O4
Formula Weight: 188.22
Melting Point: 17-19°
Boiling Point: 185-186°/18mm
MDL No.: MFCD00004427
Vzorec:
C₉H₁₆O₄ Bod varu: 185…186 °C (18 mmHg) Bod tání: 17…19 °C Hustota: 1,04 Storage Temperature: Ambient |
MDL Number:
MFCD00004427 CAS číslo: 3946-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...