Benzofurazan-5-boronic acid pinacol ester 97%
Dodavatel: Thermo Scientific
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]oxadiazole
H29358.MD
H29358.06
H29358.03
H29358.MDEA
1920
CZK
H29358.MD
H29358.06
H29358.03
Benzofurazan-5-boronic acid pinacol ester 97%
Benzofurazan-5-boronic acid pinacol ester
Purity: 97%
CAS No.: 1073355-14-2
Molecular Formula: C12H15BN2O3
Formula Weight: 246.07
Melting Point: 87-88°
MDL No.: MFCD09842717
Vzorec:
C₁₂H₁₅BN₂O₃ MW: 246,07 g/mol Bod tání: 87…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD09842717 CAS číslo: 1073355-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...