Tisk…

2-(Boc-amino)benzeneboronic acid pinacol ester 97%

Dodavatel: Thermo Scientific

Synonyms: 2-(Boc-amino)phenylboronic Acid Pinacol Ester

H29020.06 H29020.03
H29020.06EA 7860 CZK
H29020.06 H29020.03
2-(Boc-amino)benzeneboronic acid pinacol ester 97%
2-(Boc-amino)benzeneboronic acid pinacol ester

Purity: 97% CAS No.: 159624-15-4 Molecular Formula: C17H26BNO4 Formula Weight: 319.21 Melting Point: 98-100° MDL No.: MFCD03411943

Vzorec: C₁₇H₂₆BNO₄
MW: 319,21 g/mol
Bod tání: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD03411943
CAS číslo: 159624-15-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR