5-(Trifluoromethyl)indole 98%
Dodavatel: Thermo Scientific
|
Varování
|
H28782.ME
H28782.MC
H28782.MEEA
10920
CZK
H28782.ME
H28782.MC
5-(Trifluoromethyl)indole 98%
5-(Trifluoromethyl)indole
Purity: 98%
CAS No.: 100846-24-0
Molecular Formula: C9H6F3N
Formula Weight: 185.15
Melting Point: 68-71°
MDL No.: MFCD03095341
Vzorec:
C₉H₆F₃N MW: 185,15 g/mol Bod tání: 68…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095341 CAS číslo: 100846-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...