Tisk…

3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 97%

Dodavatel: Thermo Fisher Scientific
Varování

H28762.06 H28762.03
H28762.06EA 8240 CZK
H28762.06 H28762.03
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 97%
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester

Purity: 97% CAS No.: 883738-27-0 Molecular Formula: C18H29BN2O2 Formula Weight: 316.25 MDL No.: MFCD09266181

Vzorec: C₁₈H₂₉BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09266181
CAS číslo: 883738-27-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR