Dimethylphenylsilanol sodium salt 97%
Dodavatel: Thermo Fisher Scientific
H28457.03
H28457.06
H28457.03EA
2770
CZK
H28457.03
H28457.06
Dimethylphenylsilanol sodium salt 97%
Dimethylphenylsilanol sodium salt
Purity: 97%
CAS No.: 7646-75-5
Molecular Formula: C8H11NaOSi
Formula Weight: 174.25
MDL No.: MFCD09266151
Vzorec:
C₈H₁₁NaOSi MW: 174.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09266151 CAS číslo: 7646-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...