Tisk…

(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 97%

Dodavatel: Thermo Fisher Scientific

H28374.MD H28374.03
H28374.MDEA 4880 CZK
H28374.MD H28374.03
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 97%
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane

Purity: 97% CAS No.: 55739-58-7 Molecular Formula: C28H28O2P2 Formula Weight: 458.47 Melting Point: 101-105° Optical Rotation: -83° (c=1 in chloroform) MDL No.: MFCD05863546

Vzorec: C₂₈H₂₈O₂P₂
Bod tání: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD05863546
CAS číslo: 55739-58-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR